Learn More
Invitrogen™ SACM1L Polyclonal Antibody

Description
This antibody is predicted to react with bovine and rat based on sequence homology.
Specifications
Specifications
| Antigen | SACM1L |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Sacm1l |
| Gene Accession No. | Q9NTJ5 |
| Gene Alias | Kiaa0851; mKIAA0851; phosphatidylinositide phosphatase SAC1; phosphatidylinositol-3-phosphatase SAC1; phosphatidylinositide phosphatase SAC1; phosphatidylinositol-4-phosphate phosphatase; polyphosphatidylinositol phosphatase; SAC1; SAC1 (suppressor of actin mutations 1, homolog)-like (S. cerevisiae); SAC1 like phosphatidylinositide phosphatase; SAC1 suppressor of actin mutations 1 like; SAC1 suppressor of actin mutations 1-like (yeast); Sac1p; SACM1L; suppressor of actin 1; suppressor of actin mutations 1-like protein |
| Gene Symbols | Sacm1l |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.