Learn More
Invitrogen™ SACM1L Polyclonal Antibody

Description
Peptide sequence: HTLQRLSNTS PEFQEMSLLE RADQRFVWNG HLLRELSAQP EVHRFALPVL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Specifications
Specifications
| Antigen | SACM1L |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | Sacm1l |
| Gene Accession No. | Q9NTJ5 |
| Gene Alias | Kiaa0851; mKIAA0851; phosphatidylinositide phosphatase SAC1; phosphatidylinositol-3-phosphatase SAC1; phosphatidylinositide phosphatase SAC1; phosphatidylinositol-4-phosphate phosphatase; polyphosphatidylinositol phosphatase; SAC1; SAC1 (suppressor of actin mutations 1, homolog)-like (S. cerevisiae); SAC1 like phosphatidylinositide phosphatase; SAC1 suppressor of actin mutations 1 like; SAC1 suppressor of actin mutations 1-like (yeast); Sac1p; SACM1L; suppressor of actin 1; suppressor of actin mutations 1-like protein |
| Gene Symbols | Sacm1l |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.