Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SACS Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501570136
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0136 Supplier Bethyl Laboratories, Inc Supplier No. A305325A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

This gene encodes the sacsin protein, which includes a UbL domain at the N-terminus, a DnaJ domain, and a HEPN domain at the C-terminus. The gene is highly expressed in the central nervous system, also found in skin, skeletal muscles and at low levels in the pancreas. This gene includes a very large exon spanning more than 12.8 kb. Mutations in this gene result in autosomal recessive spastic ataxia of Charlevoix-Saguenay (ARSACS), a neurodegenerative disorder characterized by early-onset cerebellar ataxia with spasticity and peripheral neuropathy. Sacsin has been found to protect against mutant ataxin-1. A pseudogene associated with this gene is located on chromosome 11. Alternative splicing of this gene results in multiple transcript variants.

Specifications

Antigen SACS
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene SACS
Gene Accession No. Q9JLC8, Q9NZJ4
Gene Alias ARSACS, DKFZp686B15167, dnaJ homolog subfamily C member 29, DNAJC29, KIAA0730, PPP1R138, protein phosphatase 1, regulatory subunit 138, spastic ataxia of Charlevoix-Saguenay (sacsin), SPAX6
Gene Symbols Sacs
Host Species Rabbit
Immunogen Between 175 and 225
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26278, 50720
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.