Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAE1 Recombinant Rabbit Monoclonal Antibody (JG35-88)
GREENER_CHOICE

Catalog No. PIMA534740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534740 Supplier Invitrogen™ Supplier No. MA534740
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: SiHa, K562, 293, A549.

Posttranslational modification of proteins by the addition of the small protein SUMO (see SUMO1), or sumoylation, regulates protein structure and intracellular localization. SAE1 and UBA2. form a heterodimer that functions as a SUMO-activating enzyme for the sumoylation of proteins (Okuma et al., 1999).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAE1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JG35-88
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene SAE1
Gene Accession No. Q9UBE0
Gene Alias 2400010M20Rik; 2610044L12Rik; activator of SUMO1; AL033372; AOS1; AW743391; D7Ertd177e; FLJ3091; HSPC140; Sae1; sentrin/SUMO-activating protein AOS1; Sua1; SUMO-1 activating enzyme E1 N subunit; SUMO1 activating enzyme subunit 1; SUMO-1 activating enzyme subunit 1; SUMO-activating enzyme subunit 1; SUMO-activating enzyme subunit 1, N-terminally processed; ubiquitin-like (sentrin) activating enzyme E1A; ubiquitin-like 1 (sentrin) activating enzyme E1A; ubiquitin-like 1 (sentrin) activating enzyme subunit 1; ubiquitin-like 1-activating enzyme E1A; ubiquitin-like protein SUMO-1 activating enzyme; Ubl1a1; Uble1a
Gene Symbols SAE1
Host Species Rabbit
Immunogen Synthetic peptide within Human SAE1 aa 201-249.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10055
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.