Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAE2 Antibody (OTI4A3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273973F
Description
SAE2 Monoclonal specifically detects SAE2 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
SAE2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
UBA2 | |
Full length recombinant protein of human SAE2 (NP_005490) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG2a |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4A3 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Anthracycline-associated resistance ARX, ARX, EC 6.3.2, EC 6.3.2.-, FLJ13058, HRIHFB2115, SAE2SUMO-1 activating enzyme subunit 2, SUMO1 activating enzyme subunit 2, SUMO-activating enzyme subunit 2, UBA2, ubiquitin-activating enzyme E1 homolog, Ubiquitin-like 1-activating enzyme E1B, ubiquitin-like modifier activating enzyme 2, UBLE1B | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
10054.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction