Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAFB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582561
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582561 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582561 Supplier Invitrogen™ Supplier No. PA582561
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHGKMISVEK AKNEPVGKKT SDKRDSDGKK EKSSNSDRST NLKRDDKCDR KDDAKKGDDG SGEKSKDQDD QKPGPSERSR ATKSGSRGTE RTVV Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 81%.

This gene encodes a DNA-binding protein which has high specificity for scaffold or matrix attachment region DNA elements (S/MAR DNA). This protein is thought to be involved in attaching the base of chromatin loops to the nuclear matrix but there is conflicting evidence as to whether this protein is a component of chromatin or a nuclear matrix protein. Scaffold attachment factors are a specific subset of nuclear matrix proteins (NMP) that specifically bind to S/MAR. The encoded protein is thought to serve as a molecular base to assemble a 'transcriptosome complex' in the vicinity of actively transcribed genes. It is involved in the regulation of heat shock protein 27 transcription, can act as an estrogen receptor co-repressor and is a candidate for breast tumorigenesis. This gene is arranged head-to-head with a similar gene whose product has the same functions. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAFB
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SAFB
Gene Accession No. Q15424
Gene Alias 3110021E02Rik; 5330423C17Rik; AU018122; D18386; DKFZp779C1727; E130307D12; glutathione S-transferase fusion protein; HAP; heat-shock protein (HSP27) estrogen response element and TATA box-binding protein; HET; Hsp27 ERE-TATA binding protein; HSP27 ERE-TATA-binding protein; HSP27 estrogen response element-TATA box-binding protein; SAB-B1; Safb; SAF-B; Safb1; SAF-B1; scaffold attachment factor B; scaffold attachment factor B1
Gene Symbols SAFB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAFB. Recombinant protein control fragment (Product #RP-91677).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6294
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.