Learn More
Invitrogen™ SAFB Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117111
Description
Positive Samples: U-87MG, DU145, NIH/3T3, Mouse brain, Mouse lung, Rat brain Immunogen sequence: MAETLSGLGD SGAAGAAALS SASSETGTRR LSDLRVIDLR AELRKRNVDS SGNKSVLMER LKKAIEDEGG NPDEIEITSE GNKKTSKRSS KGRKPEEEGV EDNGLEENSG DGQEDVETSL ENLQDIDIMD ISVLDEAEID NGSVADCVED DDADNLQESL SDSRELVEGE MKELPEQLQE HAIEDKETIN NLDTSSSDFT.
This gene encodes a DNA-binding protein which has high specificity for scaffold or matrix attachment region DNA elements (S/MAR DNA). This protein is thought to be involved in attaching the base of chromatin loops to the nuclear matrix but there is conflicting evidence as to whether this protein is a component of chromatin or a nuclear matrix protein. Scaffold attachment factors are a specific subset of nuclear matrix proteins (NMP) that specifically bind to S/MAR. The encoded protein is thought to serve as a molecular base to assemble a 'transcriptosome complex' in the vicinity of actively transcribed genes. It is involved in the regulation of heat shock protein 27 transcription, can act as an estrogen receptor co-repressor and is a candidate for breast tumorigenesis. This gene is arranged head-to-head with a similar gene whose product has the same functions. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
SAFB | |
Polyclonal | |
Unconjugated | |
SAFB | |
3110021E02Rik; 5330423C17Rik; AU018122; D18386; DKFZp779C1727; E130307D12; glutathione S-transferase fusion protein; HAP; heat-shock protein (HSP27) estrogen response element and TATA box-binding protein; HET; Hsp27 ERE-TATA binding protein; HSP27 ERE-TATA-binding protein; HSP27 estrogen response element-TATA box-binding protein; SAB-B1; Safb; SAF-B; Safb1; SAF-B1; scaffold attachment factor B; scaffold attachment factor B1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
224903, 6294, 64196 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D3YXK2, O88453, Q15424 | |
SAFB | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human SAFB (NP_0011882671). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.