Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAFB2 Rabbit anti-Human, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 116421AP150UL

Description
SAF-B2 (scaffold attachment factor B2) is a nuclear matrix-associated protein that binds to matrix- or scaffold-associating regions (MARs or SARs) on DNA and interacts with RNA polymerase II and serine-/arginine-rich RNA processing factors (SR proteins). SAF-B2, and the related SAF-B, bind to the estrogen receptor (ER) and are expressed in several breast cancer cell lines at varying levels. Subsequently, SAF-B and SAF-B2 may play a role in breast cancer by mediating cellular proliferation and division. Unlike SAF-B, which is exclusively nuclear, SAF-B2 is found in the cytoplasm as well as the nucleus.Specifications
SAFB2 | |
Polyclonal | |
Unconjugated | |
SAFB2 | |
KIAA0138, SAF B2, SAFB2, scaffold attachment factor B2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
301126, 9667 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14151 | |
SAFB2 | |
SAFB2 Fusion Protein Ag2230 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction