Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aqua Phoenix™ Safe Racer 2.0 Deluxe Kit Components
Shop All Fisherbrand Products

Click to view available options
Product Type:
Alnico magnet
Battery holder
Binder Clip
D battery
Elastic cord
Nail
Plastic egg
Description
Variety of components specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits.
Specifications
Specifications
| Grade | K to 5 |
| For Use With (Application) | Education Kit |
| Length (English) | 3 in. |
| Product Type | Alnico magnet |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction