Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medisca Safe-Sense™ Gloves, 12 in.
Shop All Medisca Products

Click to view available options
Size:
Small
Medium
Large
X-Large
Description
- They are resistant to puncture abrasion and snag.
- These gloves are also very commonly used for non-hazardous compounding.
Specifications
Specifications
Accelerator Free | Yes |
Applications | Healthcare, Laboratory |
AQL | 1.5 |
Color | Blue |
Disposable | Single-use |
Exterior Finish | Fingertip Textured |
Latex Free | Yes |
Material | Nitrile |
Powdered | Powder Free |
Product Type | Nitrile Examination Glove |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction