Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAM Scientific Safe-T-Fill™ Capillary Blood Collection Systems: Serum
Click to view available options
Capacity (Metric):
200 μL
300 μL
Additive:
Clot Activator
Clot Activator and Gel
Color:
Amber
Clear
Description
Includes
Pre-assembled capillary blood collection device with a capillary tube, attached pierceable cap, and a microtube
Blood samples are drawn into the SAFE-T-FILL™ Plastic Micro Capillary Blood Collection Tubes through capillary action, reducing hemolysis, tissue contamination, and re-draws. No scooping or milking required. Coated with Sodium Fluoride for glucose determinations.
- 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
- Blood samples are drawn into the microtube through capillary action, reducing hemolysis, tissue contamination, and re-draws. No "scooping" or "milking" required
- Capillary straw and microtube are coated with anticoagulant Lithium Heparin (Green cap) to prevent clotting
- Attached cap is color-coded to designate the type of additive that is in both the capillary tube and the microtube
- Lot Number, Expiration Date, and fill lines are printed directly on the microtube
- A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
- For plasma determinations in chemistry
- Optional gel separator serves as a barrier between blood and serum after centrifugation
- Available in a variety of sizes and draw volumes, with or without gel separator
- Optional amber microtube ensures light-sensitive sample integrity providing protection for testing (ex. Bilirubin)
Recommended Storage
Store at room temperature. Keep dry. Keep away from sunlight.
Specifications
Specifications
Color | Clear |
Product Type | Chemistry/Serum Tube |
Additive | Clot Activator and Gel |
Material | Plastic |
Closure Type | Standard Pierceable Cap |
Capacity (Metric) | 200 μL |
Closure Included | Yes |
Closure Color | Red |
For Use With (Application) | Intended for chemistry determinations that require serum or clotted blood samples. Gel separator serves as a barrier between blood and serum after centrifugation. |
Description | Serum Gel Capillary Collection |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction