Learn More
ICU Medical Safe-T-Lance™ Safety Lancets
Click to view available options
Product Type:
High Flow
Low Flow
Maximum Flow Blade
Micro Flow
Normal Flow
Pediatric Blade
Color:
Blue
Green
Light Blue
Pink
Purple
Yellow
Description
- Retractable safety technology for capillary blood sampling
- Automatic retraction reduces needlestick injuries and cross-contamination exposure
- Provides a quick, gentle puncture
- Needle is shielded before and after use
- Available in six different sizes for multiple types of applications
- Lancets are color coded for easy reference
Specifications
Specifications
Disposable | Single-use |
Depth (English) Needle | Built-in Control |
Color | Green |
Needle Gauge | 21 ga |
Product Type | Normal Flow |
Quantity | 200/Box |