Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ SafeBasics™ Shoe Covers

Catalog No. p-8772037
Click to view available options
Size:
Regular
X-Large
Sole Type:
Non-skid
Standard
This item is not returnable. View return policy
This item is not returnable. View return policy

Shoe covers intended to be worn during various medical or healthcare-related procedures to protect patients and materials.

  • Slip-on
  • Sewn seams provide increased tear strength
  • Elastic opening for easy on-and-off convenience
  • Non-conductive
  • Non-Skid and regular sole options
  • 100 Shoe Covers/Box, 3 Boxes/Case, 300 Shoe Covers/Case

Specifications

Size Regular
Material Non-woven Spunbond Polypropylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Non-skid
Model 8007
Seam Type Sewn
For Use With (Application) Animal Health, Dental, Health and Wellness, Industrial, Laboratory and Research, Medical
Packaging Quantity 300/Cs.
Product Type Shoe Covers
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.