Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories SafeCup™ III 11-Panel Urine Drug Test

Catalog No. 23111442
Change view
Click to view available options
Quantity:
25/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-442 25/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-442 Supplier Germaine™ Laboratories Supplier No. 41011
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes 25 cups per case. Gloves and security seals are also included. Rapid qualitative tests for the detection of drugs of abuse in urine. Tests for AMP, BAR, BZO, BUP, COC, MTD, MDMA, MET, MOP, OXY, and PCP. CLIA Waived. Results in 5 minutes. Temperature strip included. Total collection, transport, and testing system.

Rapid qualitative tests for the detection of drugs of abuse in urine.

  • Tests for amphetamine, barbiturates, benzodiazpines, buprenorphine, cocaine, methadone, mdma, methamphetamine, morphine, oxycodone, and phencyclidine.
  • 25 cups per case.
  • CLIA Waived.
  • The total collection, testing, and transport system.
  • Easier to use, easier to read.
  • Quick results in 5 minutes.
  • Includes click seal in cap for improved leak resistance.
  • Temperature strip included.
  • Store at room temperature.
  • Adheres to SAMHSA cutoff levels.
  • Includes gloves and security seals.

Specifications

CE Marker No
Clia Complexity Waived
Format Drug Cup
Sample Type Urine
Sensitivity High
Type Drug Cup
Disposable Single-use
For Use With (Application) DOA Testing
Includes 25/Cs., Gloves, Security Seals, Package Insert
Quantity 25/Cs.
Test Time Results in 5 Minutes
Form Ready-to-Use (RTU)
Show More Show Less

CLIA Waived for OTC and POC use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.