Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDC Healthcare™ SafeGuard™ Direct Thermal Slide Labels
Click to view available options
Width (Metric):
2.22 cm
2.38 cm
Description
- Direct thermal synthetic slide labels with chemical-resistant material for histology and medical applications
- Designed to prevent barcode degradation and adhesive oozing, ensuring accurate specimen identification
- Resistant to harsh chemicals (xylene, hexane, staining reagents, alcohol, acetone, etc.)
- No need for overlay or etching systems; quick and easy printing with direct thermal printers
- Compatible with Epic and other leading EHR systems
- Packaged as 1,800 labels per roll, with 4 rolls per box
Specifications
Specifications
Adhesive Type | Permanent (Rubber-based) |
Coating | None |
Color | White |
Finish | Gloss |
Format | Label |
Material | Plastic |
Printable Sides | Single-side Printable |
Print Technology | Direct Thermal |
Product Type | Direct Thermal Printable Label |
Sterility | Non-sterile |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction