Learn More
Medicom™ SafeMask™ Premier Elite™ ProShield Earloop Mask with Visor
ASTM Level 3 mask with integrated anti-fog, distortion-free visor, latex-free, adjustable nosepiece and soft, strong earloops. Premium materials will not lint, tear or shred.
Supplier: Medicom™ 204215
This item is not returnable.
View return policy
Description
- Integrated wraparound shield protects the eyes; ideal for treating high-risk patients and for procedures producing heavy amounts of airborne particles, moisture, spray and spatter
- Non-woven spunbond outer layer provides maximum protection against fluid penetration.
- Soft, strong, ultrasonically sealed earloops attached to outside of mask prevent pulling and irritation and allow easy removal with less risk of self-contamination.
- Shingle pleat design prevents fluid pooling and cross-contamination.
- High-quality media ensures optimal filtration and breathability.
- Adjustable, enclosed nosepiece forms a strong seal to provide customized protection and comfort.
Specifications
Medical/Procedural Face Mask | |
Blue | |
Non-sterile | |
Single-use |
ASTM Level 3 | |
Snaps | |
Pleated with Eyeshield | |
Yes |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov