Learn More
Medicom™ SafeMask™ Premier Elite™ ProShield Earloop Mask with Visor
Catalog No. p-7224539
Click to view available options
Color:
Blue
Pink
Teal
Fastening Type:
Earloops
Snaps
This item is not returnable. View return policy
This item is not returnable. View return policy
ASTM Level 3 mask with integrated anti-fog, distortion-free visor, latex-free, adjustable nosepiece and soft, strong earloops. Premium materials will not lint, tear or shred.
  • Integrated wraparound shield protects the eyes; ideal for treating high-risk patients and for procedures producing heavy amounts of airborne particles, moisture, spray and spatter
  • Non-woven spunbond outer layer provides maximum protection against fluid penetration.
  • Soft, strong, ultrasonically sealed earloops attached to outside of mask prevent pulling and irritation and allow easy removal with less risk of self-contamination.
  • Shingle pleat design prevents fluid pooling and cross-contamination.
  • High-quality media ensures optimal filtration and breathability.
  • Adjustable, enclosed nosepiece forms a strong seal to provide customized protection and comfort.
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 3
Color Teal
Fastening Type Earloops
Sterility Non-sterile
Style Pleated with Eyeshield
Disposable Single-use
Latex Free Yes
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.