Learn More
Vileda Professional™ SafePlus™ System Replacement Parts
Shop All Vileda Professional Products
Click to view available options
Product Type:
Feeder for Dispenser
Flexible Lid for Dispenser
Description
- Flexible lid is antibacterial, and self-closing to ensure the dispenser is never left open
- Antibacterial properties of the lid are certified by a 3rd party, Hohenstein Labs
- Lid remains antibacterial even after 80x autoclave cycles (Certified by 3rd party, Hohensteins Labs)
- Sealed polybags wipes are packaged to fit perfectly into dispensers providing extra safety in preparation, use, and storage
Specifications
Specifications
| Product Type | Flexible Lid for Dispenser |
| Material | Silicon Rubber |
| Description | Replacement Part |
| For Use With (Equipment) | SafePlus™ Systems |
| Sterility | Non-sterile |