Learn More
Vileda Professional™ SafePlus™ System Replacement Parts
Catalog No. p-7087638 Shop All Vileda Professional Products
Change view
Click to view available options
Product Type:
Feeder for Dispenser
Flexible Lid for Dispenser
Catalog No. Product Type
18-564-2595 Feeder for Dispenser
18-564-2596 Flexible Lid for Dispenser
2 options
This item is not returnable. View return policy
Catalog No. 18-564-2595
Supplier: Vileda Professional™ 166931
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Trust the plus! The SafePlus™ System is the safe way to dispense single use microfiber wipes
  • Flexible lid is antibacterial, and self-closing to ensure the dispenser is never left open
  • Antibacterial properties of the lid are certified by a 3rd party, Hohenstein Labs
  • Lid remains antibacterial even after 80x autoclave cycles (Certified by 3rd party, Hohensteins Labs)
  • Sealed polybags wipes are packaged to fit perfectly into dispensers providing extra safety in preparation, use, and storage
Specifications
Product Type Feeder for Dispenser
Material Polypropylene
Description Replacement Part
For Use With (Equipment) SafePlus™ Systems
Sterility Non-sterile