Learn More
Safety Goggles
SCI_ED
Catalog No. s98484
Change view
Click to view available options
:
Safety Goggles
Catalog No.
S98484 Safety Goggles
1 options
This item is not returnable. View return policy
Catalog No. S98484
Supplier: Learning Resources, Inc LER2450
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
With durable plastic offering good protection to the eyes.
  • Flexible body increases wearing comfort
  • Great for kids and adults
Order Info
Grades K+.
Compliance
Meets ANSI™ Z87.1 standards
Specifications
Lens Coating Uncoated
Certifications/Compliance ANSI Z87.1
Lens Tint Clear
Product Type Safety Goggles
Material Plastic