Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Safety Multi-Sample Needles
Shop All Fisherbrand Products
Click to view available options
Gauge:
21 G
22 G
Length (English):
1.25 in.
Preattached Holder:
No
Yes
Description
- Low-angle bevel design allows easy penetration
- Color-coded caps and hubs allow easy identification of needle gauge size
- Available with safety mechanism covering the needle after use to guard against accidental needlestick during normal handling
Specifications
Specifications
| Sterility | Sterile |
| Disposable | Single-use |
| For Use With (Equipment) | Fisherbrand Blood Collection Tube Holder |
| Gauge | 21 G |
| Latex Free | Yes |
| Length (English) | 1.25 in. |
| Preattached Holder | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction