Learn More
American Educational Products Safety Pipet Filler
Click to view available options
:
Safety Pipet Filler
Description
- Made of rubber
- Three valves: one to release air, second to draw in liquid and third valve for regulating and releasing liquid
Specifications
Specifications
Color | Red |
Material | Rubber |
Type | Safety Pipette Filler |