Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius SafetySpace™ Extended Filter Tip
GREENER_CHOICE

Catalog No. 14558865 Shop All Sartorius Products
Change view
Click to view available options
Volume (Metric):
0.5 to 200 μL
10 to 1000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-558-865 0.5 to 200 μL
14-558-868 10 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-558-865 Supplier Sartorius Supplier No. LHXF780201
Add to Cart
Add to Cart

SafetySpace™ tips have extra space between the sample and the filter, which protects the sample and the pipette from cross-contamination.

SafetySpace™ Filter Tips leave more space between the sample and the filter than conventional filter tips. This allows pipetting any type of liquid or using any pipetting technique without the risk of the precious sample absorbing into the filter. In effect, this secures 100% sample recovery and thus greater accuracy.

The safety space feature is particularly useful in the following applications:

  • Pipetting foaming liquids such as buffers and proteins
  • Pipetting solvents
  • Multiple dispensing functions of electronic pipettes
  • Reverse pipetting
  • Any other application requiring extra space in the tip

The tips are made of virgin polypropylene and the filter of polyethylene without self-sealing additives to avoid any interference with the sample and the results. The filter barrier helps protect against cross-contamination of the sample and/or the pipette and reduces the need for pipette maintenance.

The filter barrier and the certified purity makes the filter tips ideal for:

  • Molecular biology
  • Microbiology
  • Cell culture applications
  • Radioactive work
  • Reduced risk of sample absorbtion into the filter with safety space feature
  • Aerosol and liquid contamination prevention with filter barrier
  • Ergonomic and light tip attachment and ejection secured by tip compatibility with the Optiload feature of Sartorius pipettes
  • Easy, single-handed opening and closing of flip-top racks
  • Easy matching with a suitable Sartorius pipette with color coded tip trays
  • Improved tip identification and traceability by informative rack labels indicating tip volume, product number, lot number, and purity level
  • Contamination free pipetting with DNase, RNase and endotoxin free certified tips
  • Environment friendly materials used in tips, trays and racks are 100% recyclable or can be incinerated as energy waste
TRUSTED_SUSTAINABILITY

Specifications

Description Free of DNase, Rnase, and endotoxins
Volume (Metric) 0.5 to 200 μL
Filtered Filtered
Sterility Sterile
Color Colorless
Autoclavable Not Autoclavable
Length (English) 3.05 in.
Length (Metric) 77.5 mm
Certifications/Compliance Manufactured according to ISO9001, ISO14001, and ISO13485; ISO class 8 clean room condition; CE complaint
For Use With (Application) Precision Liquid Dispensing
Sterilization Method E-Beam Pre-Sterilized
Format Rack
Tip Style Extended Length
Packaging Single Tray
Product Type Tips
Disposable Single-use
For Use With (Equipment) Sartorius
Graduated No
Product Line SafetySpace
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.