Learn More
Medicom™ SafeWear™ Form-Fit Isolation Gowns
Catalog No. 17227999
Click to view available options
Size:
Large
X-Large
Color:
Blue
Yellow
SafeWear™ Form-Fit Isolation Gowns are intended to protect healthcare professionals and patients from cross-infection. They are single-use and disposable.
  • Value pack of 10+2 units.
  • Professional and practical design with open back, knitted cuffs, along with a neck and waist tie for the perfect fit.
  • Latex-free.
Specifications
No. of Pockets None
Size X-Large
Gender Unisex
Product Type Isolation Gown
Material Spunbond Polypropylene
Color Blue
Closure Type Neck Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Knit
Latex Free Yes
Sleeve Style Long Sleeve
Autoclavable Not Autoclavable
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.