Learn More
Medicom™ SafeWear™ SMS Hipster Jacket Lab Coat
Catalog No. p-7224541
Click to view available options
Size:
Small
Medium
Large
X-Large
Color:
Pink
This item is not returnable. View return policy
This item is not returnable. View return policy
Procedure SMS Jacket designed to protect health professionals and patients from cross contamination. Single use and disposable gowns made of polypropylene. Material meets AAMI/PB70 Level 2 requirements for impact penetration and hydrostatic pressure. Latex free
  • High quality ultra-soft & light weight SMS fabric (Weight 45 g/m2 for Jacket & Lab Coat)
  • Fluid-resistant material meets AAMI Level 2
  • Designed to fit properly with knitted cuffs & collar
  • High-quality front snap closure buttons
  • Available in a wide range of sizes and in fashionable colors
  • 12 units in every pack
Specifications
Product Type General Purpose Lab Coat
Size Small
Gender Unisex
Material Polypropylene
Color Pink
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Style Hipster Jacket
No. of Pockets 3
Latex Free Yes
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.