Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Safranin O
SCI_ED

Catalog No. s25514 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy
Biological Stain

This product is an Educational Grade Chemical — no C of A is available.

Specifications

CAS 477-73-6
Molecular Formula C20H19ClN4
Linear Formula C20H49ClN4
MDL Number MFCD00011759
Synonym Basic Red 2
InChI Key QRYAEWIQIBAZOJ-UHFFFAOYSA-N
SMILES [Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1
IUPAC Name 2,7-diamino-1,8-dimethyl-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Molecular Weight (g/mol) 350.85
PubChem CID 2723800
Formula Weight 350.85
Packaging Solid
Color Brown/Red
Quantity 25 g
Chemical Name or Material Safranin O
Show More Show Less
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.