Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Safranin O (Certified Biological Stain), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Specifications
Specifications
Bio. Stain Comm. Certification | Pass Test |
Dye Content | Report % |
Formula Weight | 350.85 |
Grade | Certified Biological Stain |
Packaging | Glass Bottle |
Color Index No. | 50240 |
Color | Brown |
Quantity | 100 g |
Physical Form | Solid |
Chemical Name or Material | Safranin O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction