Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Safranin O (Certified Biological Stain), Fisher Chemical™

Catalog No. p-22385 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

For use in histology, cytology, bacteriology (staining)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 477-73-6
Molecular Formula C20H19ClN4
Molecular Weight (g/mol) 350.85
MDL Number MFCD00011759
InChI Key QRYAEWIQIBAZOJ-UHFFFAOYSA-N
Synonym Basic Red 2
PubChem CID 2723800
IUPAC Name 2,7-diamino-1,8-dimethyl-5-phenyl-5λ⁵-phenazin-5-ylium chloride
SMILES [Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1

Specifications

Bio. Stain Comm. Certification Pass Test
Dye Content Report %
Formula Weight 350.85
Grade Certified Biological Stain
Packaging Glass Bottle
Color Index No. 50240
Color Brown
Quantity 25 g
Physical Form Solid
Chemical Name or Material Safranin O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.