Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Safranine, Aqueous-Alcoholic, Counterstain for Gram Staining, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
Specifications
Specifications
Chemical Name or Material | Safranine |
Name Note | Counterstain for Gram Staining |
CAS | 7732-18-5, 64-17-5, 67-56-1, 447-73-6 |
CAS Min % | 90.0, 7.24, 0.32 |
CAS Max % | 94.0, 7.53, 0.33 |
Grade | Laboratory |
Packaging | Natural Poly Bottle |
Solvent or Matrix | Aqueous-Alcoholic |
Color | Dark Red |
Quantity | 120 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction