Learn More
Description
Includes
One dispenser for each reagent (A, B, and preparation Reagent) and one reagent holding rack
- Dispenser set for each chosen endpoint
- Allows for accurate testing, every time
- For use with SafTest™ kits
Specifications
Specifications
| For Use With (Application) | Alkenal Testing |
| Storage Requirements | Room temperature |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.