Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SafTest™ Sample Consumables Platform

Catalog No. p-7078526 Shop All MP Biomedicals, Inc Products
Click to view available options
Sample Type:
Liquid Samples
Solid Samples

Catalog No. MP07LC2000

Supplier: MP Biomedicals™ 07LC2000

Only null left
Add to Cart
Add to Cart

Includes

Printer paper, membranes, precision tissue wipers, solid glass beads, 15mL conical tubes, 10 to 100μL pipette tips, 200 to 1000μL pipette tips, 12mm culture tubes, 12mm culture tube caps, sharpie marke

SafTest™ Sample Consumables Platform contains all consumables necessary to run alkenal, peroxide tests.

  • Consumable platform for conducting alkenal, peroxide testing
  • Contains all of consumables necessary to run solid, liquid samples
  • Research area: Lipid degradation and oxidation
  • Detection method: Bichromatic photometer
  • Application area: Oil/fat degradation
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Peroxide, Alkenal Testing
Sample Type Liquid Samples
Quantity 1 Ea.
Storage Requirements Room temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.