Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAGE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551860
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551860 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551860 Supplier Invitrogen™ Supplier No. PA551860
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSVREEKMES GKPQTDKVIS NDAPQLGHMA AGGIPSMSTK DLYATVTQNV HEERMENNQP QPSYDLSTVL PGLTYLTVAG IPAMSTRDQY ATVTHNVHEE KIKNGQAASD NVFSTVPPAF INMAATGVSS MSTRDQYAAV THNIR Highest antigen sequence identity to the following orthologs: Mouse - 19%, Rat - 19%.

This gene belongs to a class of genes that are activated in tumors. These genes are expressed in tumors of different histologic types but not in normal tissues, except for spermatogenic cells and, for some, placenta. The proteins encoded by these genes appear to be strictly tumor specific, and hence may be excellent sources of antigens for cancer immunotherapy. This gene is expressed in sarcomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAGE1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SAGE1
Gene Accession No. Q9NXZ1
Gene Alias Cancer/testis antigen 14; CT14; putative tumor antigen; SAGE; SAGE1; Sarcoma antigen 1
Gene Symbols SAGE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAGE1. Recombinant protein control fragment (Product #RP-92247).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55511
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.