Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SAHA
GREENER_CHOICESDP

Catalog No. 465250 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-525-0 50 mg
46-5210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-525-0 Supplier R&D Systems™ Supplier No. 4652/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Class I and II HDAC inhibitor

SAHA inhibits Class I and II histone deacetylases (HDACs); induces accumulation of acetylated histones H2A, H2B, H3 and H4 in transformed cultured cells. Suppresses cell growth in a range of cancer cell lines; induces apoptosis in cutaneous T cell lymphoma cells in vitro. Activates autophagy. SAHA increases efficiency of transcription factor-induced reprogramming of mouse embryonic fibroblasts (MEF) to induced pluripotent stem cells (iPSC). Also enhances adeno-associated virus transduction of HeLa cells.

Chemical Identifiers

CAS 149647-78-9
Molecular Formula C14H20N2O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 264.32
Purity >98%
Molecular Weight (g/mol) 264.325
Chemical Name or Material SAHA
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.