Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAHH Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986280
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6280 Supplier Bioss Supplier No. BS5029R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

S-adenosylhomocysteine hydrolase (AHCY) catalyzes the reversible hydrolysis of S-adenosylhomocysteine (AdoHcy) to adenosine (Ado) and L-homocysteine (Hcy). Thus, it regulates the intracellular S-adenosylhomocysteine (SAH) concentration thought to be important for transmethylation reactions. Deficiency in this protein is one of the different causes of hypermethioninemia. S-adenosylhomocysteine hydrolase belongs to the adenosylhomocysteinase family.

Specifications

Antigen SAHH
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AHCY
Gene Accession No. P23526
Gene Alias AA987153; Acetylation; Adenosylhomocysteinase; AdoHcyase; AHCY; AL024110; BOS_13353; cb1079; CUBP; D150; dtp; ducttrip; hm:zeh1173; hm:zeh1364; Hydrolase; Liver copper-binding protein; LOC100347222; NAD; Polymorphism; S-adenosylhomocysteine hydrolase; S-adenosyl-L-homocysteine hydrolase; SAHH; wu:fj67b02
Gene Symbols AHCY
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SAHH.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 191
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.