Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SAK 3
GREENER_CHOICESDP

Catalog No. 62395 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-395 5 mg
62-392-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-395 Supplier Tocris Bioscience™ Supplier No. 6239/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent CaV3.1/3.3 activator; orally bioavailable

SAK 3 is a potent CaV3.1 and CaV3.3 activator. Enhances CaV3.1 and CaV3.3, but not CaV3.2 activity at concentrations of 0.01 - 10 M in neuro2A cells in vitro. Elevates ACh levels in hippocampus. Rescues impaired memory-related behaviors, promotes adult neurogenesis and displays antidepressant-like effects in OBX mice. Orally bioavailable.

Specifications

Chemical Name or Material Ethyl 8′-methyl-2′,4-dioxo-2-(piperidin-1-yl)-2′H-spiro[cyclopent[2]ene-1,3′-imidazo[1,2-a]pyridine]-3-carboxylate
CAS 1256269-87-0
Quantity 5 mg
Target Cav3.x Channel Activators
Molecular Formula C20H23N3O4
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.