Learn More
Description
The improved brightness and tighter binding of SALAD1 to Mango aptamers allow detection and tracking of RNA with greater sensitivity and spatial resolution, facilitating studies of RNA localization, dynamics, and expression in live-cell and single-molecule imaging applications.
Used for: imaging RNA in live and fixed cells, even at low concentration.
Application: confocal cellular imaging.
Properties and Photophysical Data: excitation and emission maxima (λ) measured in Mango selection buffer are 511 nm and 540 nm, respectively; quantum yield QY = 0.722; extinction coefficient ε(complex) = 47,284 M-1cm-1; fluorescence enhancement FE = 1,500; fluorescence efficiency E = 2,170 nM-1; brightness = 34,139 M-1cm-1. SALAD1 is cell-permeable.
Specifications
Specifications
| Chemical Name or Material | (E)-3-(2-((2-Fluoro-6-(1H-pyrazol-1-yl)benzyl)amino)-2-oxoethyl)-2-((1-methylquinolin-4(1H)-ylidene)methyl)benzo[d]thiazol-3-ium trifluoroacetate |
| Quantity | 500 μg |
| Product Line | Tocris |
| Target | Aptamers |
| Molecular Formula | C30H25FN5OS.C2F3O2 |
| Purity | 95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
