Learn More
Description
Specifications
Specifications
| Antigen | Salbutamol |
| Applications | ELISA, Lateral Flow |
| Classification | Monoclonal |
| Clone | HM531 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Salbutamol conjugated with KLH |
| Quantity | 1 mg |
| Primary or Secondary | Primary |
| Content And Storage | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.