Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Salbutamol Recombinant Rabbit Monoclonal Antibody (Huam003-82-2)
GREENER_CHOICE

Catalog No. PIMA545006
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545006 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA545006 Supplier Invitrogen™ Supplier No. MA545006
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Application notes: Indirect ELISA.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Salbutamol
Applications ELISA
Classification Recombinant Monoclonal
Clone Huam003-82-2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS, TBS with 0.05% sodium azide; pH 7.4
Host Species Rabbit
Immunogen Salbutamol linked to BSA.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.