Learn More
Invitrogen™ SALF Monoclonal Antibody (1F3)

Description
Peptide Sequence: SLKSVVVVQG AYVELQAFVN MASLAQRSSY AGSLRSCDNI RIHFPVPSQW IKALWTMNLQ RQKSLKAKMN RRACLGSLQE LESEPVIQVT VG.
Specifications
Specifications
| Antigen | SALF |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1F3 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | STON1 |
| Gene Accession No. | Q9Y6Q2 |
| Gene Alias | 4921524J06Rik; C86231; Salf; Sblf; STN1; STNB1; STON1; STON1-GTF2A1L; STON1-GTF2A1L protein; STON1-GTF2A1L readthrough; stoned B homolog 1; stoned B/TFIIA-alpha/beta-like factor; stoned B-like factor; stoned-b1; stonin 1; Stonin1; stonin-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.