Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SALF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552190 Supplier Invitrogen™ Supplier No. PA552190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSPLAISGGE SSLLPTRPTC LSHALLPSDH SCTHPTPKVG LPDEVNPQQA ESLGFQSDDL PQFQYFREDC AFSSPFWKDE GSDSHFTLDP PGSKKMFSSR NKEMPIDQKS LNKCSLNYIC EKLEHLQSAE NQDSLR Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 54%.

Endocytosis of cell surface proteins is mediated by a complex molecular machinery that assembles on the inner surface of the plasma membrane. This gene encodes one of two human homologs of the Drosophila melanogaster stoned B protein. This protein is related to components of the endocytic machinery and exhibits a modular structure consisting of an N-terminal proline-rich domain, a central region of homology specific to the human stoned B-like proteins, and a C-terminal region homologous to the mu subunits of adaptor protein (AP) complexes. Co-transcription of this gene and the neighboring downstream gene generates a rare transcript (SALF), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SALF
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STON1
Gene Accession No. Q9Y6Q2
Gene Alias 4921524J06Rik; C86231; Salf; Sblf; STN1; STNB1; STON1; STON1-GTF2A1L; STON1-GTF2A1L protein; STON1-GTF2A1L readthrough; stoned B homolog 1; stoned B/TFIIA-alpha/beta-like factor; stoned B-like factor; stoned-b1; stonin 1; Stonin1; stonin-1
Gene Symbols STON1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SALF. Recombinant protein control fragment (Product #RP-100923).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11037
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.