Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylaldehyde, 99%

Catalog No. AAA138330E
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 90-02-8 | C7H6O2 | 122.12 g/mol

Precursor to a variety of chelating agents.Salicylaldehyde is used as a precursor to prepare catechol, benzofuran, salicylaldehydimine and 3-carbethoxycoumarin. It is used for colorimetric assay of isoleucine as well as utilized for the determination of primary amine by fluorescence method. It is a key starting material to a various chelating agents. It is used as flavor and fragrance components. Its derivative salicylaldehyde thiosemicarbazone is used for the simultaneous determination of molybdenum and iron by spectrophotometric method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Precursor to a variety of chelating agents.Salicylaldehyde is used as a precursor to prepare catechol, benzofuran, salicylaldehydimine and 3-carbethoxycoumarin. It is used for colorimetric assay of isoleucine as well as utilized for the determination of primary amine by fluorescence method. It is a key starting material to a various chelating agents. It is used as flavor and fragrance components. Its derivative salicylaldehyde thiosemicarbazone is used for the simultaneous determination of molybdenum and iron by spectrophotometric method.

Solubility
Miscible with alcohol, ether, acetone, benzene and most organic solvents. Slightly miscible with water.

Notes
Incompatible with acids, strong reducing agents, strong oxidizing agents, strong bases and halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-02-8
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00003317
InChI Key SMQUZDBALVYZAC-UHFFFAOYSA-N
Synonym salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
PubChem CID 6998
ChEBI CHEBI:16008
IUPAC Name 2-hydroxybenzaldehyde
SMILES OC1=CC=CC=C1C=O

Specifications

Melting Point -7°C
Density 1.168
Boiling Point 197°C
Flash Point 76°C (168°F)
Odor Almond-like
Assay Percent Range 99%
Refractive Index 1.573
Quantity 2500 g
Beilstein 471388
Sensitivity Air and light sensitive
Merck Index 14,8326
Solubility Information Miscible with alcohol,ether,acetone,benzene and most organic solvents. Slightly miscible with water.
Formula Weight 122.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Salicylaldehyde
Show More Show Less
Hazard Category H227-H302-H311-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P309+P311-P312-P314-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 201-961-0
RTECSNumber VN5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.