Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salicylaldehyde, Analytical Standard, ≥98.5% (GC), MilliporeSigma™ Supelco™
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mL
Specifications
Specifications
CAS | 90-02-8 |
Vapor Pressure | 1 mmHg (33°C) |
Vapor Density | 4.2 (vs. air) |
Density | 1.146 g/mL (at 25°C) |
Linear Formula | 2-(HO)C6H4CHO |
MDL Number | MFCD00003317 |
Melting Point | 1°C to 2°C |
Boiling Point | 197°C |
Synonym | 2-Hydroxybenzaldehyde |
Quantity | 1 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction