Learn More
Salicylaldehyde azine, 97%
Description
Salicylaldehyde azine is used in the preparation of 2-oxo-2H-1-benzopyran-3-carboxylic acid 2-(2-oxo-2H-1-benzopyran-3-carbonyl)hydrazide by reacting with propadienedione . It has anthelmintic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Salicylaldehyde Azine is used in the preparation of 2-oxo-2H-1-benzopyran-3-carboxylic acid 2-(2-oxo-2H-1-benzopyran-3-carbonyl)hydrazide by reacting with propadienedione . It has anthelmintic activity.
Solubility
Soluble in dimethyl sulfoxide, methanol and ethyl acetate. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 216°C to 222°C |
Assay Percent Range | 97% |
Quantity | 10 g |
Beilstein | 1913597 |
Solubility Information | Soluble in dimethyl sulfoxide,methanol and ethyl acetate. Slightly soluble in water. |
Formula Weight | 240.26 |
Percent Purity | 97% |
Chemical Name or Material | Salicylaldehyde Azine |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 213-500-0 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.