Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylaldoxime, 98%

Catalog No. p-7081207
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 94-67-7 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-67-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00002120
InChI Key GFCNBJDXQMZOOC-WAYWQWQTSA-N
Synonym salicylaldoxime, 2-hydroxybenzaldehyde oxime, salicylaldehyde oxime, 2-hydroxyimino methyl phenol, 6-hydroxyamino methylidene cyclohexa-2,4-dien-1-one, e-2-hydroxybenzaldehyde oxime, orihzizptztncu-uhfffaoysa-n, orihzizptztncu-vmpitwqzsa-n, 2-1e-hydroxyimino methyl phenol, salicylideneaminoalcohol
PubChem CID 6740756
IUPAC Name 6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
SMILES ON\C=C1\C=CC=CC1=O

Specifications

Melting Point 55°C to 59°C
Quantity 25 g
Beilstein 1859881
Merck Index 14,8327
Formula Weight 137.14
Percent Purity 98%
Chemical Name or Material Salicylaldoxime
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-353-8
RTECSNumber VN5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.