Learn More
Salicylamide, 98%
Description
Salicylamide is used in combination with both aspirin and caffeine in the over-the-counter pain remedies. It is used as an analgesic and as an antipyretic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Salicylamide is used in combination with both aspirin and caffeine in the over-the-counter pain remedies. It is used as an analgesic and as an antipyretic.
Solubility
Soluble in water (2.06 mg/ml at 25°C), chloroform, dimethyl sulfoxide, alcohol, and methanol (100 mg/ml).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 138°C to 142°C |
Boiling Point | 181°C (14 mmHg) |
Quantity | 250 g |
Beilstein | 742439 |
Merck Index | 14,8328 |
Solubility Information | Soluble in water (2.06mg/ml at 25°C),chloroform,dimethyl sulfoxide,alcohol,and methanol (100mg/ml). |
Formula Weight | 137.14 |
Percent Purity | 98% |
Chemical Name or Material | Salicylamide |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 200-609-3 |
RTECSNumber | VN6475000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.