Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylhydroxamic acid, 99%, Thermo Scientific Chemicals

Catalog No. AC132620250
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

Catalog No. AC132620250

Supplier: Thermo Scientific Chemicals 132620250

Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-73-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
InChI Key HBROZNQEVUILML-UHFFFAOYSA-N
Synonym salicylhydroxamic acid, 2-hydroxybenzohydroxamic acid, salicylohydroxamic acid, sham, salicylohydroximic acid, benzamide, n,2-dihydroxy, 2-hydroxybenzhydroxamic acid, o-hydroxybenzohydroxamic acid, sha, 2,n-dihydroxy-benzamide
PubChem CID 66644
ChEBI CHEBI:45615
IUPAC Name N,2-dihydroxybenzamide
SMILES C1=CC=C(C(=C1)C(=O)NO)O

Specifications

Melting Point 177.0°C to 185.0°C
Color Beige-Pink
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H4C(=NOH)OH
Quantity 25 g
Beilstein 10,98
Merck Index 15,8468
Solubility Information Solubility in water: slightly soluble. Other solubilities: very soluble in ethanol, diethyl ether, soluble with alkalies
Formula Weight 153.14
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Salicylhydroxamic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 201-934-3
RTECSNumber VO6870000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.