Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylic acid, ACS, 99+%

Catalog No. AA3078236
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 69-72-7 | C7H6O3 | 138.12 g/mol

Salicylic acid is used as a food preservative as well as used in the treatment of many skin disorders such as acne, dandruff, psoriasis and seborrheic dermatitis of the skin and scalp. It is an antiseptic. It is mainly used in the pharmaceutical industry for the manufacture of antipyretic, analgesic, anti-inflammatory and diuretic drugs. It is also used in the dye industry for making azo direct dyes and acidic mordant dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-72-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002439
InChI Key YGSDEFSMJLZEOE-UHFFFAOYSA-N
Synonym salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
PubChem CID 338
ChEBI CHEBI:16914
IUPAC Name 2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC=C1O

Specifications

Melting Point 158°C to 160°C
Boiling Point 211°C (20mmHg)
Beilstein 774890
Merck Index 14,8332
Solubility Information Soluble in carbon terachloride,benzene,propanol,absolute ethanol,acetone,toluene and ether; Slightly soluble in cold water; More soluble in boiling water; Water solubility increased by Na3PO4,alkali acetates or alkali citrates.
Formula Weight 138.12
Percent Purity ≥99%
Grade ACS
Density 1.443 g/mL
Quantity 500 g
Flash Point 157°C (315°F)
Physical Form Powder
Chemical Name or Material Salicylic acid
Show More Show Less
Hazard Category H302-H318-H361d
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P305+P351+P338-P308+P313-P310-P330-P501c
EINECSNumber 200-712-3
RTECSNumber VO0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.