Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salicylic Acid, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical

Click to view available options
Quantity:
125 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Salicylic Acid, Crystal, Reagent, ACS is an alpha-hydroxybenzoic acid and a colorless crystalline acid that is organic and commonly used in organic synthesis. It can be found occurring naturally in unripe fruits and vegetables and most commonly in the white willow tree. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Specifications
Specifications
Melting Point | 158°C to 161°C |
CAS Max % | 1 |
Percent Purity | 99% |
Grade | ACS |
Quantity | 12 kg |
Packaging | Poly Pail |
Residue after Ignition | 0.0001 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction