Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salicylideneaniline, 97%, Thermo Scientific™
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Specifications
Specifications
| Melting Point | 49.4°C to 52.4°C |
| Color | Tan to White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | C6H5NCHC6H4(OH) |
| Quantity | 25 g |
| Solubility Information | Solubility in water: insoluble in water. Other solubilities: soluble in acetone, dichloromethane, chloroform, slightly soluble in hexane, methanol |
| Formula Weight | 197.24 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction