Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Salinomycin Monoclonal Antibody (4F7)
GREENER_CHOICE

Catalog No. PIMA549621
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549621 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549621 Supplier Invitrogen™ Supplier No. MA549621
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Salinomycin is a polyether antibiotic that has been found to have potent anticancer properties, particularly against cancer stem cells. It was first isolated from Streptomyces albus in the late 1970s and works by binding to and inhibiting the activity of ATP synthase, a protein involved in cellular energy production. This disruption of energy production leads to cell death in cancer cells, while sparing normal cells. Salinomycin also has antibacterial and antifungal properties, making it a promising candidate for the development of new antibiotics.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Salinomycin
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 4F7
Concentration 5.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias Coxistac; Procoxacin
Host Species Mouse
Immunogen Conjugate of SLM hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.